Thiazole-2-carbaldehyde ≥97.0%
Supplier: TCI America
F0303-5G
F0303-1G
TCF0303-5GEA
202.24
CAD
TCF0303-5G
TCF0303-001G
Thiazole-2-carbaldehyde ≥97.0%
Thiazole-2-carbaldehyde
CAS Number: 10200-59-6
MDL Number: MFCD00142924
Molecular Formula: C4H3NOS
Molecular Weight: 113.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Flash Point (°C): 68
Specific Gravity (20/20): 1.30
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2-Thiazolecarboxaldehyde
Formula:
C₄H₃NOS Boiling Pt: 70 °C Density: 1.30 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00142924 CAS Number: 10200-59-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...