1-(2-Fluorophenyl)piperazine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

F0291-25G F0291-5G
TCF0291-025GEA 566.9 CAD
TCF0291-025G TCF0291-5G
1-(2-Fluorophenyl)piperazine hydrochloride ≥98.0% (by titrimetric analysis)
1-(2-Fluorophenyl)piperazine hydrochloride

CAS Number: 1011-16-1
MDL Number: MFCD00134409
Molecular Formula: C10H13FN2
Molecular Weight: 216.68
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 190

Formula: C₁₀H₁₃FN₂
Melting Pt: 190 °C
MDL Number: MFCD00134409
CAS Number: 1011-16-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR