Ferroceneboronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)

Supplier: TCI America

F0280-1G F0280-100MG
TCF0280-001GEA 406.06 CAD
TCF0280-001G TCF0280-100MG
Ferroceneboronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
Ferroceneboronic acid

[Cyclic boronating reagent for GC/MS]
CAS Number: 12152-94-2
MDL Number: MFCD00059074
Molecular Formula: C10H11BFeO2
Molecular Weight: 229.85
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal

Formula: C₁₀H₁₁BFeO₂
MDL Number: MFCD00059074
CAS Number: 12152-94-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edd55c2a6805f8edcac87b55911d6744c77ef72f&vid=edd55c2a6805f8edcac87b55911d6744c77ef72f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR