3-(2-Fluorophenoxy)benzyl bromide

Supplier: TCI America

Synonyms: 3'-Bromomethyl-2-fluorodiphenyl Ether

F0236-1G F0236-100MG
TCF0236-001GEA 742.13 CAD
TCF0236-001G TCF0236-100MG
3-(2-Fluorophenoxy)benzyl bromide
3-(2-Fluorophenoxy)benzyl bromide

CAS Number: 242812-04-0
MDL Number: MFCD00059914
Molecular Formula: C13H10BrFO
Molecular Weight: 281.12
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 1.46
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3'-Bromomethyl-2-fluorodiphenyl Ether

Formula: C₁₃H₁₀BrFO
MW: 281.12 g/mol
Density: 1.46 (20 °C)
MDL Number: MFCD00059914
CAS Number: 242812-04-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a4a603e77f2e72df0fb5a4c42458d379f494a12&vid=8a4a603e77f2e72df0fb5a4c42458d379f494a12&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR