2-Fluoro-β-alanine hydrochloride ≥99.0%

Supplier: TCI America

Synonyms: 3-Amino-2-fluoropropionic Acid Hydrochloride

F0185-1G F0185-100MG
TCF0185-1GEA 635.16 CAD
TCF0185-1G TCF0185-100MG
2-Fluoro-β-alanine hydrochloride ≥99.0%
2-Fluoro-beta-alanine hydrochloride

CAS Number: 867-84-5
MDL Number: MFCD00060202
Molecular Formula: C3H6FNO2
Molecular Weight: 143.54
Purity/Analysis Method: >99.0% (N)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Amino-2-fluoropropionic Acid Hydrochloride

Formula: C₃H₆FNO₂
MW: 143.54 g/mol
MDL Number: MFCD00060202
CAS Number: 867-84-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0a1c6e01b0f843553b2c302d1f8aeb4ef707100&vid=a0a1c6e01b0f843553b2c302d1f8aeb4ef707100&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR