4-Fluoro-N,N-dimethylaniline ≥98.0%

Supplier: TCI America

F0175-5G F0175-1G
TCF0175-5GEA 358.66 CAD
TCF0175-5G TCF0175-001G
4-Fluoro-N,N-dimethylaniline ≥98.0%
4-Fluoro-N,N-dimethylaniline

CAS Number: 403-46-3
MDL Number: MFCD00059471
Molecular Formula: C8H10FN
Molecular Weight: 139.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 36

Formula: C₈H₁₀FN
Boiling Pt: 200 °C
Melting Pt: 36 °C
MDL Number: MFCD00059471
CAS Number: 403-46-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cca8ca3c41dad506bb1e14b34f62e8f3ef17bd8c&vid=cca8ca3c41dad506bb1e14b34f62e8f3ef17bd8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR