N-Formyl-L-tyrosine ≥98.0%

Supplier: TCI America

F0135-1G F0135-5G
TCF0135-001GEA 306.52 CAD
TCF0135-001G TCF0135-005G
N-Formyl-L-tyrosine ≥98.0%
N-Formyl-L-tyrosine

CAS Number: 13200-86-7
MDL Number: MFCD00021031
Molecular Formula: C10H11NO4
Molecular Weight: 209.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 165
Specific rotation [a]20/D: 83 deg (C=2.6, EtOH)

Formula: C₁₀H₁₁NO₄
MW: 209.20 g/mol
Melting Pt: 165 °C
MDL Number: MFCD00021031
CAS Number: 13200-86-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=653f6ce0fcef9e36ad2cc9a632591b495f52d507&vid=653f6ce0fcef9e36ad2cc9a632591b495f52d507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR