N-Formyl-L-leucine
Supplier: TCI America
Synonyms:
N-Formyl-Leu-OH, For-Leu-OH
F0131-5G
F0131-1G
TCF0131-5GEA
135.56
CAD
TCF0131-5G
TCF0131-001G
N-Formyl-L-leucine
N-Formyl-L-leucine
CAS Number: 6113-61-7
MDL Number: MFCD00055861
Molecular Formula: C7H13NO3
Molecular Weight: 159.19
Form: Crystal
Color: White
Melting point (°C): 142
Specific rotation [a]20/D: -39.5 deg (C=1, H2O)
Formula:
C₇H₁₃NO₃ Melting Pt: 142 °C |
MDL Number:
MFCD00055861 CAS Number: 6113-61-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...