N-Formyl-DL-phenylalanine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

F0126-25G F0126-5G
TCF0126-025GEA 378.57 CAD
TCF0126-025G TCF0126-005G
N-Formyl-DL-phenylalanine ≥99.0% (by titrimetric analysis)
N-Formyl-DL-phenylalanine

CAS Number: 4289-95-6
MDL Number: MFCD00063281
Molecular Formula: C10H11NO3
Molecular Weight: 193.20
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 167

Formula: C₁₀H₁₁NO₃
Melting Pt: 167 °C
MDL Number: MFCD00063281
CAS Number: 4289-95-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e529c4a5ea80d5ee196b01ce021fb8d9c4dfbf1b&vid=e529c4a5ea80d5ee196b01ce021fb8d9c4dfbf1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR