N-Formyl-D-phenylalanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

F0125-100MG F0125-1G
TCF0125-100MGEA 45.82 CAD
TCF0125-100MG TCF0125-001G
N-Formyl-D-phenylalanine ≥98.0% (by titrimetric analysis)
N-Formyl-D-phenylalanine

CAS Number: 59366-89-1
MDL Number: MFCD00063282
Molecular Formula: C10H11NO3
Molecular Weight: 193.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 165
Specific rotation [a]20/D: -66 deg (C=4, EtOH)

Formula: C₁₀H₁₁NO₃
Melting Pt: 165 °C
MDL Number: MFCD00063282
CAS Number: 59366-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fec6c9255bb793e91a285d432f23ec09da352e6&vid=3fec6c9255bb793e91a285d432f23ec09da352e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR