N-Formyl-DL-ethionine 98,0

Supplier: TCI America

F0120-1G F0120-100MG
TCF0120-001GEA 576.86 CAD
TCF0120-001G TCF0120-100MG
N-Formyl-DL-ethionine 98,0
N-Formyl-DL-ethionine

MDL Number: MFCD00059815
Molecular Formula: C7H13NO3S
Molecular Weight: 191.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 117

Formula: C₇H₁₃NO₃S
Melting Pt: 117 °C
MDL Number: MFCD00059815
CAS Number: 126872-00-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffc87ab95a1b3c1a1d4b5c42bfc413973c215cc8&vid=ffc87ab95a1b3c1a1d4b5c42bfc413973c215cc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR