Dibutyl Fumarate ≥98.0%

Supplier: TCI America

Synonyms: Dibutyl fumarate, Fumaric Acid Dibutyl Ester

F0116-25G F0116-500G
TCF0116-025GEA 61.62 CAD
TCF0116-025G TCF0116-500G
Dibutyl Fumarate ≥98.0%
Dibutyl Fumarate

CAS Number: 105-75-9
MDL Number: MFCD00065141
Molecular Formula: C12H20O4
Molecular Weight: 228.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 285
Melting point (°C): -18
Flash Point (°C): 142
Specific Gravity (20/20): 0.99


Product Categories: Materials Chemistry, Resin Additives, Copolymerization Monomers
Other Product Names / Synonyms: Fumaric Acid Dibutyl Ester

Formula: C₁₂H₂₀O₄
MW: 228.29 g/mol
Boiling Pt: 285 °C
Melting Pt: –18 °C
Density: 0.99 (20 °C)
Flash Pt: 142 °C
MDL Number: MFCD00065141
CAS Number: 105-75-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9292d483db517536e61f5edd81373564ec9dda5a&vid=9292d483db517536e61f5edd81373564ec9dda5a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR