Formamidine hydrochloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Formimidamide hydrochloride

F0103-25G F0103-5G
TCF0103-025GEA 230.68 CAD
TCF0103-025G TCF0103-005G
Formamidine hydrochloride ≥97.0% (by titrimetric analysis)
Formamidine hydrochloride

CAS Number: 6313-33-3
MDL Number: MFCD00012865
Molecular Formula: CH4N2
Molecular Weight: 80.52
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 85

Formula: CH₄N₂
MW: 80.52 g/mol
Melting Pt: 85 °C
MDL Number: MFCD00012865
CAS Number: 6313-33-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f865cf6a8dfe84e7b8cdfa708083f322d69dbe85&vid=f865cf6a8dfe84e7b8cdfa708083f322d69dbe85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR