ɑ-Furil dioxime ≥97.0% (by gravimetric analysis)
Supplier: TCI America
F0079-5G
F0079-1G
TCF0079-5GEA
2333.66
CAD
TCF0079-5G
TCF0079-001G
ɑ-Furil dioxime ≥97.0% (by gravimetric analysis)
alpha-Furil dioxime
CAS Number: 23789-34-6
MDL Number: MFCD00075315
Molecular Formula: C10H8N2O4
Molecular Weight: 220.18
Purity/Analysis Method: >97.0% (W)
Form: Crystal
Melting point (°C): 192
Formula:
C₁₀H₈N₂O₄ Melting Pt: 192 °C |
MDL Number:
MFCD00075315 CAS Number: 23789-34-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...