Formoxime 10% ∼2.4 mol/L in water

Supplier: TCI America

Synonyms: Formaldoxime

F0044-25ML F0044-500ML
TCF0044-025MLEA 61.07 CAD
TCF0044-025ML TCF0044-500ML
Formoxime 10% ∼2.4 mol/L in water
Formoxime

CAS Number: 75-17-2
MDL Number: MFCD00058969
Molecular Formula: CH3NO
Molecular Weight: 45.04
Form: Clear Liquid
Color: Colorless
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Formaldoxime

Formula: CH₃NO
MW: 45.04 g/mol
MDL Number: MFCD00058969
CAS Number: 75-17-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e388edea9e72200a721b83386dd31f256f2d108b&vid=c67b17cb5f542547a5769b8cf24be4ca5a08a633&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR