Fluorescin ≥95.0%

Supplier: TCI America

F0028-5G F0028-25G
TCF0028-5GEA 369.72 CAD
TCF0028-5G TCF0028-025G
Fluorescin ≥95.0%
Fluorescin

[Reagent for Oxydases and Peroxydases]
CAS Number: 518-44-5
MDL Number: MFCD00046931
Molecular Formula: C20H14O5
Molecular Weight: 334.33
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 127

Formula: C₂₀H₁₄O₅
Melting Pt: 127 °C
MDL Number: MFCD00046931
CAS Number: 518-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce28a4a4b058d186eb82f3c27057bbe0bc1c0d77&vid=ce28a4a4b058d186eb82f3c27057bbe0bc1c0d77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR