2-Acetyl-7-hydroxybenzofuran ≥98.0%

Supplier: TCI America

Synonyms: 1-(7-Hydroxybenzofuran-2-yl)ethan-1-one, 2-Acetyl-7-hydroxycoumarone

A1316-5G A1316-10G
TCA1316-5GEA 143.78 CAD
TCA1316-5G TCA1316-010G
2-Acetyl-7-hydroxybenzofuran ≥98.0%
2-Acetyl-7-hydroxybenzofuran

CAS Number: 40020-87-9
MDL Number: MFCD00191373
Molecular Formula: C10H8O3
Molecular Weight: 176.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 166


Other Product Names / Synonyms: 2-Acetyl-7-hydroxycoumarone

Formula: C₁₀H₈O₃
Melting Pt: 166 °C
MDL Number: MFCD00191373
CAS Number: 40020-87-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c64b359dfe2d440e0d4e6f4f9b9461623b71272b&vid=c64b359dfe2d440e0d4e6f4f9b9461623b71272b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR