Ethyl-7-chloro-2-oxoheptanoate ≥98.0%
Supplier: TCI America
Synonyms:
7-Chloro-2-oxoheptanoic Acid Ethyl Ester, Ethyl 7-chloro-2-oxoheptanoate
E0827-25G
E0827-5G
TCE0827-25GEA
287.56
CAD
TCE0827-25G
TCE0827-5G
Ethyl-7-chloro-2-oxoheptanoate ≥98.0%
Ethyl-7-chloro-2-oxoheptanoate
CAS Number: 78834-75-0
MDL Number: MFCD07698675
Molecular Formula: C9H15ClO3
Molecular Weight: 206.67
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.11
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 7-Chloro-2-oxoheptanoic Acid Ethyl Ester
Formula:
C₉H₁₅ClO₃ Density: 1.11 (20 °C) |
MDL Number:
MFCD07698675 CAS Number: 78834-75-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...