Ethanolamine thioglycolate ∼85% in water

Supplier: TCI America

Synonyms: Ethanolamine Mercaptoacetate, Monoethanolamine Thioglycolate

E0813-25G E0813-500G
TCE0813-25GEA 36.34 CAD
TCE0813-25G TCE0813-500G
Ethanolamine thioglycolate ∼85% in water
Ethanolamine thioglycolate

CAS Number: 126-97-6
MDL Number: MFCD00050789
Molecular Formula: C4H11NO3S
Molecular Weight: 153.20
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: Monoethanolamine Thioglycolate, Ethanolamine Mercaptoacetate

Formula: C₄H₁₁NO₃S
MW: 153.20 g/mol
MDL Number: MFCD00050789
CAS Number: 126-97-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=023bf9a0d8ace80c8b3809614e10218863d4e04f&vid=f6f84e2f4e725a2f71d0da4efc9eb3dc6e36cf20&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR