Ethyl-DL-leucate ≥98.0%

Supplier: TCI America

Synonyms: DL-Leucic Acid Ethyl Ester, Ethyl DL-2-Hydroxy-4-methylvalerate, DL-2-Hydroxy-4-methylvaleric Acid Ethyl Ester

E0802-25G E0802-5G
TCE0802-25GEA 717.32 CAD
TCE0802-25G TCE0802-5G
Ethyl-DL-leucate ≥98.0%
Ethyl-DL-leucate

CAS Number: 10348-47-7
Molecular Formula: C8H16O3
Molecular Weight: 160.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Specific Gravity (20/20): 0.97
Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: DL-Leucic Acid Ethyl Ester, Ethyl DL-2-Hydroxy-4-methylvalerate, DL-2-Hydroxy-4-methylvaleric Acid Ethyl Ester

Formula: C₈H₁₆O₃
MW: 160.21 g/mol
Boiling Pt: 185 °C
Density: 0.97 (20 °C)
CAS Number: 10348-47-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6f287c363451bec14b3d6839e40ac9ed5035cd0&vid=e6f287c363451bec14b3d6839e40ac9ed5035cd0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR