5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone ≥97.0% (by GC, titration analysis)

Supplier: TCI America

E0800-5G E0800-25G
TCE0800-5GEA 93.25 CAD
TCE0800-5G TCE0800-25G
5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone ≥97.0% (by GC, titration analysis)
5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone

CAS Number: 698-10-2
MDL Number: MFCD00036673
Molecular Formula: C7H10O3
Molecular Weight: 142.15
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 94
Flash Point (°C): 113
Freezing point (°C): 27


Other Product Names / Synonyms: Ethyl Fenugreek Lactone

Formula: C₇H₁₀O₃
Boiling Pt: 94 °C
Flash Pt: 113 °C
MDL Number: MFCD00036673
CAS Number: 698-10-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a18b45240fd38495f1768162fd18ba374ad2f2b&vid=3a18b45240fd38495f1768162fd18ba374ad2f2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR