N-Ethyl-2-isopropyl-5-methylcyclohexanecarboxamide ≥98.0%

Supplier: TCI America

Synonyms: TRPM8 agonist WS-3, N-Ethyl-p-menthane-3-carboxamide, Ethyl menthane carboxamide

E0796-5G E0796-25G
TCE0796-5GEA 31.6 CAD
TCE0796-5G TCE0796-25G
N-Ethyl-2-isopropyl-5-methylcyclohexanecarboxamide ≥98.0%
N-Ethyl-2-isopropyl-5-methylcyclohexanecarboxamide

CAS Number: 39711-79-0
MDL Number: MFCD00130071
Molecular Formula: C13H25NO
Molecular Weight: 211.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 99
Specific rotation [a]20/D: -52 deg (C=1, MeOH)


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: N-Ethyl-2-isopropyl-5-methylcyclohexanecarboxamide

Formula: C₁₃H₂₅NO
MW: 211.349 g/mol
Melting Pt: 99 °C
MDL Number: MFCD00130071
CAS Number: 39711-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7865865c264135e9c525ae4133be0bbeded068a7&vid=7865865c264135e9c525ae4133be0bbeded068a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR