3-Ethyl-2-hydroxy-2-cyclopenten-1-one ≥97.0%

Supplier: TCI America

E0792-25G E0792-5G
TCE0792-25GEA 180.12 CAD
TCE0792-25G TCE0792-5G
3-Ethyl-2-hydroxy-2-cyclopenten-1-one ≥97.0%
3-Ethyl-2-hydroxy-2-cyclopenten-1-one

CAS Number: 21835-01-8
MDL Number: MFCD00012391
Molecular Formula: C7H10O2
Molecular Weight: 126.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 75
Melting point (°C): 42
Flash Point (°C): 107
Storage Temperature: <0°C

Formula: C₇H₁₀O₂
Boiling Pt: 75 °C
Melting Pt: 42 °C
Flash Pt: 107 °C
MDL Number: MFCD00012391
CAS Number: 21835-01-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=774b3723cf2e68528326734be2ae744233888bd9&vid=774b3723cf2e68528326734be2ae744233888bd9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR