4'-Methoxyacetoacetanilide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N-(4-Methoxyphenyl)-3-oxobutanamide

A1307-25G A1307-500G
TCA1307-025GEA 51.19 CAD
TCA1307-025G TCA1307-500G
4'-Methoxyacetoacetanilide ≥98.0% (by HPLC, total nitrogen)
4'-Methoxyacetoacetanilide

CAS Number: 5437-98-9
MDL Number: MFCD00008783
Molecular Formula: C11H13NO3
Molecular Weight: 207.23
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 117


Other Product Names / Synonyms: N-Acetoacetyl-p-anisidine, N-Acetoacetyl-4-methoxyaniline

Formula: C₁₁H₁₃NO₃
MW: 207.2300 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00008783
CAS Number: 5437-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23838bbe2dcfe6a2c8a2794633fec815c45382e6&vid=23838bbe2dcfe6a2c8a2794633fec815c45382e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR