3-Carbethoxy-2-piperidone ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 2-Piperidone-3-carboxylate, 2-Oxo-3-piperidinecarboxylic acid ethyl ester, Ethyl 2-Oxo-3-piperidinecarboxylate, 3-Ethoxycarbonyl-2-piperidone

E0742-25G E0742-5G
TCE0742-25GEA 240.22 CAD
TCE0742-25G TCE0742-5G
3-Carbethoxy-2-piperidone ≥98.0%
3-Carbethoxy-2-piperidone

CAS Number: 3731-16-6
MDL Number: MFCD00006038
Molecular Formula: C8H13NO3
Molecular Weight: 171.20
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 82


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Ethyl 2-Piperidone-3-carboxylate, Ethyl 2-Oxo-3-piperidinecarboxylate, 2-Oxo-3-piperidinecarboxylic Acid Ethyl Ester

Formula: C8H13NO3
MW: 171.1900 g/mol
Melting Pt: 82 °C
MDL Number: MFCD00006038
CAS Number: 3731-16-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbbcde83771d2bb557ba9c3c20e2f2e436d8c9d7&vid=dbbcde83771d2bb557ba9c3c20e2f2e436d8c9d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR