Ethyl-6-bromocoumarin-3-carboxylate ≥98.0%

Supplier: TCI America

E0733-5G E0733-25G
TCE0733-5GEA 183.44 CAD
TCE0733-5G TCE0733-25G
Ethyl-6-bromocoumarin-3-carboxylate ≥98.0%
Ethyl-6-bromocoumarin-3-carboxylate

CAS Number: 2199-90-8
MDL Number: MFCD00124584
Molecular Formula: C12H9BrO4
Molecular Weight: 297.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 164


Other Product Names / Synonyms: 6-Bromocoumarin-3-carboxylic Acid Ethyl Ester

Formula: C₁₂H₉BrO₄
Melting Pt: 164 °C
MDL Number: MFCD00124584
CAS Number: 2199-90-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=170093900b2f0bab4d3b55267b95d95b9e19a88d&vid=170093900b2f0bab4d3b55267b95d95b9e19a88d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR