2-(4-Ethoxyphenyl)-2-methylpropanol ≥98.0%
Supplier: TCI America
E0730-25G
E0730-5G
TCE0730-25GEA
244.58
CAD
TCE0730-25G
TCE0730-5G
2-(4-Ethoxyphenyl)-2-methylpropanol ≥98.0%
2-(4-Ethoxyphenyl)-2-methylpropanol
CAS Number: 83493-63-4
MDL Number: MFCD07787279
Molecular Formula: C12H18O2
Molecular Weight: 194.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 48
Formula:
C₁₂H₁₈O₂ Melting Pt: 48 °C |
MDL Number:
MFCD07787279 CAS Number: 83493-63-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...