2-Ethyl-3-(methylthio)pyrazine ≥98.0%

Supplier: TCI America

E0719-25G E0719-5G
TCE0719-25GEA 358.66 CAD
TCE0719-25G TCE0719-5G
2-Ethyl-3-(methylthio)pyrazine ≥98.0%
2-Ethyl-3-(methylthio)pyrazine

CAS Number: 72987-62-3
MDL Number: MFCD00038027
Molecular Formula: C7H10N2S
Molecular Weight: 154.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: 2-(Methylthio)-3-ethylpyrazine

Formula: C₇H₁₀N₂S
Density: 1.11 (20 °C)
MDL Number: MFCD00038027
CAS Number: 72987-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ff59fac4d5a83d323ff7abc68d6dcece399cf92&vid=3ff59fac4d5a83d323ff7abc68d6dcece399cf92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR