4,5-Dimethyl-2-ethylthiazole ≥98.0%

Supplier: TCI America

E0718-5G E0718-25G
TCE0718-5GEA 50.56 CAD
TCE0718-5G TCE0718-25G
4,5-Dimethyl-2-ethylthiazole ≥98.0%
4,5-Dimethyl-2-ethylthiazole

CAS Number: 873-64-3
MDL Number: MFCD00053125
Molecular Formula: C7H11NS
Molecular Weight: 141.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Flash Point (°C): 74
Specific Gravity (20/20): 1.01

Formula: C₇H₁₁NS
Boiling Pt: 185 °C
Density: 1.01 (20 °C)
Flash Pt: 74 °C
MDL Number: MFCD00053125
CAS Number: 873-64-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa254077edccc01ac0f5b65622b902eeb0dfca8a&vid=fa254077edccc01ac0f5b65622b902eeb0dfca8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR