2'-Methoxyacetoacetanilide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A1306-25G A1306-500G
TCA1306-025GEA 26.86 CAD
TCA1306-025G TCA1306-500G
2'-Methoxyacetoacetanilide ≥98.0% (by HPLC, total nitrogen)
2'-Methoxyacetoacetanilide

CAS Number: 92-15-9
MDL Number: MFCD00008781
Molecular Formula: C11H13NO3
Molecular Weight: 207.23
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 85
Flash Point (°C): 163


Other Product Names / Synonyms: N-Acetoacetyl-o-anisidine, N-Acetoacetyl-2-methoxyaniline

Formula: C₁₁H₁₃NO₃
Melting Pt: 85 °C
Flash Pt: 163 °C
MDL Number: MFCD00008781
CAS Number: 92-15-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ccdf58491ebcae871b84e371ed0ae8719e9de2a&vid=0ccdf58491ebcae871b84e371ed0ae8719e9de2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR