2-Ethylpyrrole ≥96.0%
Supplier: TCI America
Synonyms:
2-Ethyl-1H-pyrrole
E0713-1G
E0713-5G
TCE0713-1GEA
122.18
CAD
TCE0713-1G
TCE0713-5G
2-Ethylpyrrole ≥96.0%
2-Ethylpyrrole
CAS Number: 1551-06-0
MDL Number: MFCD00060477
Molecular Formula: C6H9N
Molecular Weight: 95.15
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 62
Specific Gravity (20/20): 0.93
Storage Temperature: 0-10°C
Formula:
C₆H₉N MW: 95.15 g/mol Boiling Pt: 165 °C Density: 0.93 (20 °C) Flash Pt: 62 °C |
MDL Number:
MFCD00060477 CAS Number: 1551-06-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...