Ethyl-5-bromo-1H-indole-2-carboxylate ≥98.0%
Supplier: TCI America
Synonyms:
Ethyl 5-bromoindole-2-carboxylate, 5-Bromoindole-2-carboxylic acid ethyl ester, Ethyl 5-Bromoindole-2-carboxylate, Ethyl 5-bromo-1H-indole-2-carboxylate
E0698-1G
E0698-5G
TCE0698-1GEA
36.34
CAD
TCE0698-1G
TCE0698-5G
Ethyl-5-bromo-1H-indole-2-carboxylate ≥98.0%
Ethyl-5-bromo-1H-indole-2-carboxylate
CAS Number: 16732-70-0
MDL Number: MFCD00022701
Molecular Formula: C11H10BrNO2
Molecular Weight: 268.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 165
Other Product Names / Synonyms: 5-Bromoindole-2-carboxylic Acid Ethyl Ester
Formula:
C₁₁H₁₀BrNO₂ MW: 268.11 g/mol Melting Pt: 165 °C |
MDL Number:
MFCD00022701 CAS Number: 16732-70-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...