1-Ethyl-3-methylpyridinium ethylsulfate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

E0681-5G E0681-25G
TCE0681-5GEA 285.98 CAD
TCE0681-5G TCE0681-25G
1-Ethyl-3-methylpyridinium ethylsulfate ≥98.0% (by HPLC, total nitrogen)
1-Ethyl-3-methylpyridinium ethyl sulphate

CAS Number: 872672-50-9
Molecular Formula: C10H17NO4S
Molecular Weight: 247.31
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Clear Liquid
Storage Temperature: 0-10°C

Formula: C₁₀H₁₇NO₄S
MW: 247.32 g/mol
CAS Number: 872672-50-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a87cd111136915b5e7f616e2db4a1532382ea5b&vid=7a87cd111136915b5e7f616e2db4a1532382ea5b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR