2-(2-Ethoxyethoxy)ethyl acrylate ≥98.0% stabilized

Supplier: TCI America

Synonyms: Di(ethylene glycol) ethyl ether acrylate, Acrylic Acid 2-(2-Ethoxyethoxy)ethyl Ester, Diethylene Glycol Monoethyl Ether Acrylate

E0652-500G E0652-25G
TCE0652-500GEA 255.41 CAD
TCE0652-500G TCE0652-25G
2-(2-Ethoxyethoxy)ethyl acrylate ≥98.0% stabilized
2-(2-Ethoxyethoxy)ethyl acrylate

(stabilized with MEHQ)
CAS Number: 7328-17-8
Molecular Formula: C9H16O4
Molecular Weight: 188.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 250
Flash Point (°C): 116
Specific Gravity (20/20): 1.02


Product Categories: Materials Chemistry, Monomers, Bioscience, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: Acrylic Acid 2-(2-Ethoxyethoxy)ethyl Ester, Diethylene Glycol Monoethyl Ether Acrylate

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 250 °C
Density: 1.02 (20 °C)
Flash Pt: 116 °C
MDL Number: MFCD00015655
CAS Number: 7328-17-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1aef554cdbc0f727dfdbacc656eeae0142f06ec1&vid=1aef554cdbc0f727dfdbacc656eeae0142f06ec1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR