3-Amino-5,5-dimethyl-2-cyclohexen-1-one ≥98.0%

Supplier: TCI America

Synonyms: 3-Amino-5,5-dimethylcyclohex-2-enone

A1300-25G A1300-5G
TCA1300-25GEA 590.13 CAD
TCA1300-25G TCA1300-005G
3-Amino-5,5-dimethyl-2-cyclohexen-1-one ≥98.0%
3-Amino-5,5-dimethyl-2-cyclohexen-1-one

CAS Number: 873-95-0
MDL Number: MFCD00045531
Molecular Formula: C8H13NO
Molecular Weight: 139.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 166

Formula: C₈H₁₃NO
Melting Pt: 166 °C
MDL Number: MFCD00045531
CAS Number: 873-95-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afad85cbbf72e66cff66710b9b8567a515bc2170&vid=afad85cbbf72e66cff66710b9b8567a515bc2170&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR