Ethyl-7-(diethylamino)coumarin-3-carboxylate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
7-(Diethylamino)coumarin-3-carboxylic Acid Ethyl Ester, Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate
CAS Number: 28705-46-6
MDL Number: MFCD00227484
Molecular Formula: C16H19NO4
Molecular Weight: 289.33
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 86
Product Categories: Materials Chemistry, Coumarin Dyes (Functional Organic Dyes), Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 7-(Diethylamino)coumarin-3-carboxylic Acid Ethyl Ester
Formula:
C₁₆H₁₉NO₄ MW: 289.33 g/mol Melting Pt: 86 °C |
MDL Number:
MFCD00227484 CAS Number: 28705-46-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...