Ethyl-4-acetamidophenylacetate ≥98.0%

Supplier: TCI America

Synonyms: 4-Acetamidophenylacetic Acid Ethyl Ester

E0585-25G E0585-5G
TCE0585-25GEA 139.04 CAD
TCE0585-25G TCE0585-5G
Ethyl-4-acetamidophenylacetate ≥98.0%
Ethyl-4-acetamidophenylacetate

CAS Number: 13475-17-7
MDL Number: MFCD06797104
Molecular Formula: C12H15NO3
Molecular Weight: 221.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 78


Other Product Names / Synonyms: 4-Acetamidophenylacetic Acid Ethyl Ester

Formula: C₁₂H₁₅NO₃
MW: 221.26 g/mol
Melting Pt: 78 °C
MDL Number: MFCD06797104
CAS Number: 13475-17-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55b1beda4d51daa809a18728a9c2c061f065d98b&vid=55b1beda4d51daa809a18728a9c2c061f065d98b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR