N-Ethyl-N-(2-hydroxyethyl)nitrosamine ≥97.0%

Supplier: TCI America

E0580-1G E0580-5G
TCE0580-1GEA 48.98 CAD
TCE0580-1G TCE0580-5G
N-Ethyl-N-(2-hydroxyethyl)nitrosamine ≥97.0%
N-Ethyl-N-(2-hydroxyethyl)nitrosamine

CAS Number: 13147-25-6
MDL Number: MFCD01727871
Molecular Formula: C4H10N2O2
Molecular Weight: 118.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Specific Gravity (20/20): 1.10
Storage Temperature: 0-10°C


Other Product Names / Synonyms: N-Ethyl-N-nitroso-2-aminoethanol

Formula: C₄H₁₀N₂O₂
Boiling Pt: 103 °C
Density: 1.10 (20 °C)
MDL Number: MFCD01727871
CAS Number: 13147-25-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b3d64309cea71b8e0ce15f8f4f0b99c8cec1486&vid=9b3d64309cea71b8e0ce15f8f4f0b99c8cec1486&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR