4-Ethynylphthalic anhydride ≥98.0%

Supplier: TCI America

E0579-1G E0579-5G
TCE0579-1GEA 381.89 CAD
TCE0579-1G TCE0579-5G
4-Ethynylphthalic anhydride ≥98.0%
4-Ethynylphthalic anhydride

CAS Number: 73819-76-8
MDL Number: MFCD06200801
Molecular Formula: C10H4O3
Molecular Weight: 172.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 125

Formula: C₁₀H₄O₃
MW: 172.14 g/mol
Melting Pt: 125 °C
MDL Number: MFCD06200801
CAS Number: 73819-76-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80a25c312e244c162d068006945264222ef39e81&vid=80a25c312e244c162d068006945264222ef39e81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR