1,4-Epoxy-1,4-dihydronaphthalene ≥98.0%

Supplier: TCI America

E0567-1G E0567-5G
TCE0567-1GEA 178.54 CAD
TCE0567-1G TCE0567-5G
1,4-Epoxy-1,4-dihydronaphthalene ≥98.0%
1,4-Epoxy-1,4-dihydronaphthalene

CAS Number: 573-57-9
MDL Number: MFCD00167584
Molecular Formula: C10H8O
Molecular Weight: 144.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 57
Flash Point (°C): 93


Other Product Names / Synonyms: 1,4-Dihydro-1,4-epoxynaphthalene

Formula: C₁₀H₈O
Melting Pt: 57 °C
Flash Pt: 93 °C
MDL Number: MFCD00167584
CAS Number: 573-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=627da87a47477ebe5996da69dfec129d2f172455&vid=627da87a47477ebe5996da69dfec129d2f172455&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR