EDTA tetrasodium salt tetrahydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

E0565-500G E0565-25G
TCE0565-500GEA 95.7 CAD
TCE0565-500G TCE0565-25G
EDTA tetrasodium salt tetrahydrate ≥98.0% (by titrimetric analysis)
EDTA tetrasodium salt tetrahydrate

CAS Number: 13235-36-4
MDL Number: MFCD00150027
Molecular Formula: C10H16N2O8
Molecular Weight: 380.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Ethylenediaminetetraacetic Acid Tetrasodium Salt

Formula: C₁₀H₁₂N₂O₈Na₄·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150027
CAS Number: 13235-36-4
EINECS: 200-573-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3092b3ce810d517cb493feee0307ed67be682f1d&vid=e9d26e0050feabcfe9b5addd81a9ba6dff51834d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR