1-Hexyl-4-ethynylbenzene ≥95.0%

Supplier: TCI America

Synonyms: 4-Hexylphenylacetylene, 1-Ethynyl-4-hexylbenzene, (4-Hexylphenyl)acetylene, 4-Hexyl-1-ethynylbenzene, (4-Hexylphenyl)ethyne

E0564-5G E0564-25G
TCE0564-5GEA 176.96 CAD
TCE0564-5G TCE0564-25G
1-Hexyl-4-ethynylbenzene ≥95.0%
1-Hexyl-4-ethynylbenzene

CAS Number: 79887-11-9
MDL Number: MFCD00173883
Molecular Formula: C14H18
Molecular Weight: 186.30
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 100
Specific Gravity (20/20): 0.89


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Hexyl-1-ethynylbenzene, 4-Hexylphenylacetylene

Formula: C₁₄H₁₈
MW: 186.3
Density: 0.89 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00173883
CAS Number: 79887-11-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc86d25dfe7a6be0f2065910fe89bffc64a1bc71&vid=fc86d25dfe7a6be0f2065910fe89bffc64a1bc71&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR