2-(Ethylamino)pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0558-25G E0558-5G
TCE0558-25GEA 532.3 CAD
TCE0558-25G TCE0558-5G
2-(Ethylamino)pyridine ≥98.0% (by GC, titration analysis)
2-(Ethylamino)pyridine

CAS Number: 37059-57-7
MDL Number: MFCD04038418
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 82
Specific Gravity (20/20): 1.02

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 82 °C
Density: 1.02 (20 °C)
MDL Number: MFCD04038418
CAS Number: 37059-57-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07a73a39c52b720fbbb31b1e013b8ce23551cd34&vid=07a73a39c52b720fbbb31b1e013b8ce23551cd34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR