2,4-Dimethyl-3-ethylpyrrole ≥97.0%
Supplier: TCI America
Synonyms:
Cryptopyrrole, 3-Ethyl-2,4-dimethylpyrrole, Kryptopyrrole
E0555-5G
E0555-1G
TCE0555-5GEA
243.29
CAD
TCE0555-5G
TCE0555-1G
2,4-Dimethyl-3-ethylpyrrole ≥97.0%
2,4-Dimethyl-3-ethylpyrrole
CAS Number: 517-22-6
MDL Number: MFCD00005222
Molecular Formula: C8H13N
Molecular Weight: 123.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 71
Specific Gravity (20/20): 0.91
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Cryptopyrrole, Kryptopyrrole
Formula:
C₈H₁₃N MW: 123.2 g/mol Density: 0.91 (20 °C) Flash Pt: 71 °C |
MDL Number:
MFCD00005222 CAS Number: 517-22-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...