3-Ethyl-3-oxetanemethanol ≥96.0% (by GC) for synthesis

Supplier: TCI America

Synonyms: (3-Ethyloxetan-3-yl)methanol, 2-Hydroxymethyl-2-ethyl-1,3-epoxypropane, 3-Hydroxymethyl-3-ethyloxetane

E0546-25G E0546-500G E0546-100G
TCE0546-25GEA 58.46 CAD
TCE0546-25G TCE0546-500G TCE0546-100G
3-Ethyl-3-oxetanemethanol ≥96.0% (by GC) for synthesis
3-Ethyl-3-oxetanemethanol

CAS Number: 3047-32-3 MDL Number: MFCD00190143 Molecular Formula: C6H12O2 Molecular Weight: 116.16 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Boiling point (°C): 110 Flash Point (°C): 112 Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: 2-Hydroxymethyl-2-ethyl-1,3-epoxypropane, 3-Hydroxymethyl-3-ethyloxetane

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 110 °C (12mmHg) (lit.)
Density: 1.02 (20 °C)
Flash Pt: 112 °C (lit.)
MDL Number: MFCD00190143
CAS Number: 3047-32-3
EINECS: 221-254-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45de692da41ef548f48425ee36d381edf09823a1&vid=45de692da41ef548f48425ee36d381edf09823a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR