3-(Ethylamino)phenol hemisulfate ≥98.0%

Supplier: TCI America

E0534-1G E0534-5G
TCE0534-1GEA 173.8 CAD
TCE0534-1G TCE0534-5G
3-(Ethylamino)phenol hemisulfate ≥98.0%
3-(Ethylamino)phenol hemisulfate

CAS Number: 1274892-48-6
MDL Number: MFCD03093630
Molecular Formula: C8H11NO
Molecular Weight: 186.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₈H₁₁NO
MW: 186.22 g/mol
MDL Number: MFCD03093630
CAS Number: 1274892-48-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1899641ce9fc39c16567812bbe5e71a291fed60&vid=a1899641ce9fc39c16567812bbe5e71a291fed60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR