Ethyl nonafluorobutyl ether (mixture of isomers) ≥98.0%

Supplier: TCI America

Synonyms: Perfluorobutyl Ethyl Ether, Nonafluorobutyl Ethyl Ether, Ethyl Perfluorobutyl Ether

E0528-25G E0528-500G
TCE0528-025GEA 67.94 CAD
TCE0528-025G TCE0528-500G
Ethyl nonafluorobutyl ether (mixture of isomers) ≥98.0%
Ethyl nonafluorobutyl ether (mixture of isomers)

CAS Number: 813458-04-7
Molecular Formula: C6H5F9O
Molecular Weight: 264.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.44


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Ethyl Perfluorobutyl Ether, Nonafluorobutyl Ethyl Ether, Perfluorobutyl Ethyl Ether

Formula: C₆H₅F₉O
MW: 264.09 g/mol
Density: 1.44 (20 °C)
CAS Number: 813458-04-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40144e747bc4e6431225f171b324598868828068&vid=40144e747bc4e6431225f171b324598868828068&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR