N-(3-Aminopropyl)-N-methylaniline ≥96.0% (by GC, titration analysis)

Supplier: TCI America

A1295-25ML A1295-500ML
TCA1295-025MLEA 125.61 CAD
TCA1295-025ML TCA1295-500ML
N-(3-Aminopropyl)-N-methylaniline ≥96.0% (by GC, titration analysis)
N-(3-Aminopropyl)-N-methylaniline

CAS Number: 53485-07-7
MDL Number: MFCD00191602
Molecular Formula: C10H16N2
Molecular Weight: 164.25
Purity/Analysis Method: >96.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 172
Specific Gravity (20/20): 1.00

Formula: C₁₀H₁₆N₂
Boiling Pt: 172 °C
Density: 1.00 (20 °C)
MDL Number: MFCD00191602
CAS Number: 53485-07-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08487b0bf560d546f1bceba606fb0d56d3f8b2d6&vid=08487b0bf560d546f1bceba606fb0d56d3f8b2d6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR