3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

E0511-5G E0511-25G
TCE0511-005GEA 52.14 CAD
TCE0511-005G TCE0511-025G
3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide ≥98.0% (by HPLC, titration analysis)
3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide

CAS Number: 54016-70-5
MDL Number: MFCD00040549
Molecular Formula: C8H14BrNOS
Molecular Weight: 252.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 84

Formula: C₈H₁₄BrNOS
Melting Pt: 84 °C
MDL Number: MFCD00040549
CAS Number: 54016-70-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce94d1150225be7c5c94ce676a4ba380c09821d7&vid=ce94d1150225be7c5c94ce676a4ba380c09821d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR