Ethylene glycol monoacetoacetate monomethacrylate ≥95.0% (by HPLC) stabilized

Supplier: TCI America

Synonyms: 2-(Methacryloyloxy)ethyl Acetoacetate, AAEM, Acetoacetic Acid 2-(Methacryloyloxy)ethyl Ester, 3-oxo-butyrate 2-[2-methyl-2-acrylyl] -oxygen ethyl ester, Lonzamon AAMEA PQ, Acetoacetic acid 2-(Methacryloyloxy)ethyl Ester, Methacrylic acid 2-(Acetoacetyloxy)ethyl Ester, Ethylene Glycol Monoacetoacetate Monomethacrylate, Methacrylic Acid 2-(Acetoacetyloxy)ethyl Ester, 2-(Acetoacetyloxy)ethyl Methacrylate

E0489-25G E0489-500G
TCE0489-025GEA 56.88 CAD
TCE0489-025G TCE0489-500G
Ethylene glycol monoacetoacetate monomethacrylate ≥95.0% (by HPLC) stabilized
Ethylene glycol monoacetoacetate monomethacrylate

(stabilized with BHT)
CAS Number: 21282-97-3
MDL Number: MFCD00054405
Molecular Formula: C10H14O5
Molecular Weight: 214.22
Purity/Analysis Method: >95.0% (HPLC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 141
Specific Gravity (20/20): 1.12


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction), Materials Chemistry, Monomers
Other Product Names / Synonyms: Acetoacetic Acid 2-(Methacryloyloxy)ethyl Ester, 2-(Acetoacetyloxy)ethyl Methacrylate, Methacrylic Acid 2-(Acetoacetyloxy)ethyl Ester, 2-(Methacryloyloxy)ethyl Acetoacetate

Formula: C₁₀H₁₄O₅
MW: 214.22 g/mol
Boiling Pt: 100 °C
Density: 1.12 (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00054405
CAS Number: 21282-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ab7048a72b732500358c2f80e88b46d51111246&vid=4ab7048a72b732500358c2f80e88b46d51111246&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR